missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Methylstyrene (stabilized with TBC) 99.0+%, TCI America™
$41.02 - $52.19
Chemical Identifiers
CAS | 98-83-9 |
---|---|
Molecular Formula | C9H10 |
Molecular Weight (g/mol) | 118.18 |
MDL Number | MFCD00008859 |
InChI Key | XYLMUPLGERFSHI-UHFFFAOYSA-N |
Synonym | alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene |
PubChem CID | 7407 |
ChEBI | CHEBI:35060 |
IUPAC Name | (prop-1-en-2-yl)benzene |
SMILES | CC(=C)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M042925ML
|
TCI America
M042925ML |
25 mL |
Each for $41.02
|
|
|||||
M0429500ML
|
TCI America
M0429500ML |
500 mL |
Each for $52.19
|
|
|||||
Chemical Identifiers
98-83-9 | |
118.18 | |
XYLMUPLGERFSHI-UHFFFAOYSA-N | |
7407 | |
(prop-1-en-2-yl)benzene |
C9H10 | |
MFCD00008859 | |
alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene | |
CHEBI:35060 | |
CC(=C)C1=CC=CC=C1 |
Specifications
98-83-9 | |
Colorless | |
C9H10 | |
25 mL | |
alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene | |
CC(=C)C1=CC=CC=C1 | |
118.18 | |
CHEBI:35060 | |
≥99.0% (GC) | |
alpha-Methylstyrene (stabilized with TBC) |
-23°C | |
162°C | |
MFCD00008859 | |
2303 | |
XYLMUPLGERFSHI-UHFFFAOYSA-N | |
(prop-1-en-2-yl)benzene | |
7407 | |
118.18 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-0005&(3)-0008
RTECSNumber : WL5075300
TSCA : Yes
Recommended Storage : Refrigerator