missing translation for 'onlineSavingsMsg'
Learn More

alpha-Hexylcinnamaldehyde 90.0+%, TCI America™

$70.84 - $175.22

Chemical Identifiers

CAS 101-86-0
Molecular Formula C15H20O
Molecular Weight (g/mol) 216.32
MDL Number MFCD00006989
InChI Key GUUHFMWKWLOQMM-NTCAYCPXSA-N
Synonym 2-Hexyl-3-phenylacrylaldehyde, 2-Hexyl-3-phenyl-2-propenal
PubChem CID 1715135
IUPAC Name (2E)-2-(phenylmethylidene)octanal
SMILES CCCCCC\C(C=O)=C/C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H068525ML
View Documents
TCI America
H068525ML
25 mL
Each for $70.84
Only null left
 
H0685500ML
View Documents
TCI America
H0685500ML
500 mL
Each for $175.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

101-86-0
216.32
GUUHFMWKWLOQMM-NTCAYCPXSA-N
1715135
CCCCCC\C(C=O)=C/C1=CC=CC=C1
C15H20O
MFCD00006989
2-Hexyl-3-phenylacrylaldehyde, 2-Hexyl-3-phenyl-2-propenal
(2E)-2-(phenylmethylidene)octanal

Specifications

101-86-0
175°C
MFCD00006989
2-Hexyl-3-phenylacrylaldehyde, 2-Hexyl-3-phenyl-2-propenal
CCCCCC\C(C=O)=C/C1=CC=CC=C1
216.32
216.32
Liquid
Yellow
C15H20O
25 mL
GUUHFMWKWLOQMM-NTCAYCPXSA-N
(2E)-2-(phenylmethylidene)octanal
1715135
≥90.0% (GC)
alpha-Hexylcinnamaldehyde
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2657

RTECSNumber : GD6560000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'