missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allantoin, Powder, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor AL21312KG
Specifications
97-59-6 | |
Poly Pail | |
0.98 |
1 | |
12 kg | |
Reagent |
Chemical Identifiers
97-59-6 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.