missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alendronic Acid 98.0+%, TCI America™
$124.89 - $370.95
Chemical Identifiers
CAS | 66376-36-1 |
---|---|
Molecular Formula | C4H13NO7P2 |
Molecular Weight (g/mol) | 249.096 |
MDL Number | MFCD00868112 |
InChI Key | OGSPWJRAVKPPFI-UHFFFAOYSA-N |
Synonym | 4-Amino-1-hydroxybutane-1,1-diphosphonic Acid |
PubChem CID | 2088 |
ChEBI | CHEBI:2567 |
IUPAC Name | (4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid |
SMILES | C(CC(O)(P(=O)(O)O)P(=O)(O)O)CN |
Chemical Identifiers
66376-36-1 | |
249.096 | |
OGSPWJRAVKPPFI-UHFFFAOYSA-N | |
2088 | |
(4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid |
C4H13NO7P2 | |
MFCD00868112 | |
4-Amino-1-hydroxybutane-1,1-diphosphonic Acid | |
CHEBI:2567 | |
C(CC(O)(P(=O)(O)O)P(=O)(O)O)CN |
Specifications
66376-36-1 | |
White-Yellow | |
MFCD00868112 | |
1759 | |
OGSPWJRAVKPPFI-UHFFFAOYSA-N | |
(4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid | |
2088 | |
249.10 | |
Crystalline Powder |
235°C | |
C4H13NO7P2 | |
5 g | |
4-Amino-1-hydroxybutane-1,1-diphosphonic Acid | |
C(CC(O)(P(=O)(O)O)P(=O)(O)O)CN | |
249.096 | |
CHEBI:2567 | |
≥98.0% (T) | |
Alendronic Acid |
Safety and Handling
RTECSNumber : SZ6521833
TSCA : No