Learn More
AirClean™ Systems TeachAide™ Demonstration Fume Hoods
Provides protection from experiments using or generating toxic fumes, vapors, particulate or smoke
Supplier:  AirClean™ Systems ACTA32
 View more versions of this product
Catalog No. 36-101-4669
Only null left
Explore available promotions
Description
Description
Protects teacher and students from exposure to hazardous chemical and particulate associated with hands-on science experiments.
  • AirSafe™ automatic safety controller display
  • Combination with airflow monitoring and gas detection
  • Sparkless/brushless motor is post-filter and deep base contains accidental spills
  • Low airflow alarm warns user both visually and audibly
  • Filtration and electronics located above work area to minimize chance of chemical spills
  • Meets OSHA/ANSI regulations and guidelines for fume hood airflow
Filtration
  • Uses revolutionary bonded carbon filter
  • Filter minimizes potential exposure to hazardous carbon dust
  • Carbon pellets evenly distributed throughout filter allow maximum capture yielding higher load capacity
AirSafe Features
  • Constant airflow monitoring and adjustment to preset velocity
  • Constant monitoring of filter condition
  • Audible and visible alarms
Optional
  • Built-in polypropylene cup sink and drain
  • Electrical cord access ports, water and gas options
  • Custom cart available in fixed 34 in. height with shelving options. (Meets ADA requirements)
  • Fluorescent lighting standard on ACTA48 and optional ACTA32
Specifications
Specifications
Ductless Fume Hood
58.4 x 78.7 x 66 cm (23 x 31 x 26 in.)
Product Suggestions
Product Suggestions