missing translation for 'onlineSavingsMsg'
Learn More

PURELL™ Advanced Hand Sanitizer Foam Refill

Foam refill (1200 mL) for PURELL™ ES10 touch-free hand sanitizer dispensers.

$126.39 - $126.39

Specifications

Volume (Metric) 1200 mL
Color Clear
View More Specs

Products 1
Catalog Number Mfr. No. Solution Type Price Quantity  
Catalog Number Mfr. No. Solution Type Price Quantity  
19-043-366
View Documents
PURELL™
835102CAN00
Hand Sanitizer Fragrance-Free Foam
Case of 2 Each for $126.39
Only null left
 
Description

Description

  • Hand sanitizer foam is made with high-quality ethyl alcohol produced in the USA.
  • Manufactured according to strict FDA safety and quality standards.
  • Formula meets USDA Certified Biobased Product Standards for hand cleaners and Cradle to Cradle Certified™.
  • 1200 mL refill for the PURELL™ ES10 Automatic Hand Sanitizer Dispenser (sold separately) bottle is made with recyclable plastic components.
  • Collar and alkaline AA battery easily pull apart making it easy to recycle and reduce product waste.
  • Case Includes: (2) PURELL™ Advanced Hand Sanitizer Foam, 1200 mL Refills for the ES10 Automatic Hand Sanitizer Dispenser.
  • ES10 dispensers (8324-E1, 8320-E1, 8328-E1) are sold separately.
  • Ultra nourishing formula is enhanced with a moisturizer and antioxidant blend.
Specifications

Specifications

1200 mL
Clear
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'