missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Adenine, ≥98%, Calbiochem™,

$122.00 - $479.00

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
115210GM
View Documents
MilliporeSigma™
115210GM
10 g Glass Bottle
Each for $122.00
Only null left
 
115225GM
View Documents
MilliporeSigma™
115225GM
25 g Glass Bottle
Each for $142.00
Only null left
 
1152100GM
View Documents
MilliporeSigma™
1152100GM
100 g Glass Bottle
Each for $479.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

73-24-5
135.13
GFFGJBXGBJISGV-UHFFFAOYSA-N
190
7H-purin-6-amine
C5H5N5
MFCD00041790
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
CHEBI:16708
NC1=C2NC=NC2=NC=N1

Specifications

73-24-5
Glass Bottle
MFCD00041790
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
NC1=C2NC=NC2=NC=N1
135.13
CHEBI:16708
Solid
White
C5H5N5
10 g
GFFGJBXGBJISGV-UHFFFAOYSA-N
7H-purin-6-amine
190
>98% by Assay; ≥99% by HPLC
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'