missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Activin A Monoclonal Antibody (KAA378_7F6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555693
Description
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.Specifications
Activin A | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P08476 | |
Inhba | |
Recombinant human INHBA (proprietary range information). | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
KAA378_7F6 | |
Unconjugated | |
Inhba | |
activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product | |
Mouse | |
Protein A | |
RUO | |
3624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |