missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Activin A Monoclonal Antibody (KAA378_4H12), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA555694

 View more versions of this product

Catalog No. MA555694


Only null left
Explore available promotions

Description

Description

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
Specifications

Specifications

Activin A
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P08476
Inhba
Recombinant human INHBA (proprietary range information).
50 μg
Primary
Human
Antibody
IgG2b
ELISA, Western Blot
KAA378_4H12
Unconjugated
Inhba
activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product
Mouse
Protein A
RUO
3624
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions