missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Powder, USP, Spectrum™ Chemical
CA191, 7440-44-0, C
$415.89 - $25900.89
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
IUPAC Name | carbon |
SMILES | [C] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18600308
|
Spectrum Chemical Mfg Cor
CA191125GM |
125 g | Amber Glass Bottle |
Each for $415.89
|
|
||||
18600309
|
Spectrum Chemical Mfg Cor
CA191500GM |
500 g | Amber Glass Bottle |
Each for $1,116.94
|
|
||||
18600310
|
Spectrum Chemical Mfg Cor
CA19125KG |
2.5 kg | Poly Pail |
Each for $3,700.49
|
|
||||
18600311
|
Spectrum Chemical Mfg Cor
CA19112KG |
12 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18600312
|
Spectrum Chemical Mfg Cor
CA19145KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Activated Charcoal, Powder, USP is carbon that has been treated to create low-volume pores that increase the area available for chemical reactions and adsorption. The most common pharmaceutical uses of activated charcoal is as a purification agent and antitoxin. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
7440-44-0 | |
12.01 | |
carbon |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
[C] |
Specifications
7440-44-0 | |
4% | |
Amber Glass Bottle | |
125 g | |
[C] | |
12.01 |
100% | |
15% | |
C | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
USP |