missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Gradient grade, 99.9% min, OmniSolv, MilliporeSigma™
$159.21 - $3034.03
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
MAX01426
|
Technidata
AX01426 |
1 L | Glass Bottle |
Each for $159.21
|
|
||||
MAX01421
|
Technidata
AX01421 |
4 L | Glass Bottle |
Each for $707.55
Case of 4 Each for $2,830.20
|
|
||||
MAX0142NP20
|
Technidata
AX0142NP20 |
20 L | NOWPak™ |
Each for $3,034.03
|
|
||||
Chemical Identifiers
75-05-8 | |
41.053 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
Specifications
75-05-8 | |
1 L | |
Liquid | |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 |
Colorless | |
Glass Bottle | |
99.9% min. | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
Acetonitrile |