missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, ≥99.9%, Honeywell Burdick & Jackson
$2761.23 - $2761.23
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6000268
|
Honeywell-Burdick and Jackson
NP01520 |
20 L | HDPE Nowpak™ |
Each for $2,761.23
|
|
||||
Chemical Identifiers
75-05-8 | |
41.053 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
Specifications
75-05-8 | |
0.7822 g/cm3 | |
20 L | |
HDPE Nowpak™ | |
UN1648 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
>99.9% | |
Acetonitrile, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, ≥99.9% |
100.0000% | |
82°C | |
6°C | |
C2H3N | |
741857 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41 | |
HPLC Gradient |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile