missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, For pesticide residue analysis, Honeywell Riedel-de Haën™
for pesticide residue analysis
Supplier: Honeywell Chemicals 344816X1L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Acetonitrile | |
Colorless | |
6 x 1 L | |
2°C (35.6°F) | |
C2H3N | |
MFCD00001878 | |
UN1648 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 |
75-05-8 | |
0.786g/mL at 25°C | |
0.16 | |
973°F | |
CH3CN | |
72.8 mmHg (20°C) | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.9% (GC) |
Chemical Identifiers
75-05-8 | |
MFCD00001878 | |
6342 | |
acetonitrile |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
CHEBI:38472 | |
CC#N |
Safety and Handling
P210-P280-P305 + P351 + P338
DOTInformation : 3 / PGII
RTECSNumber : 200-835-2