missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile 70% and Water 30%, LabReady™ Solvent Blend, Honeywell Burdick & Jackson
$1303.38 - $1303.38
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Chemical Identifiers
75-05-8 | |
41.053 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
Specifications
75-05-8,7732-18-5 | |
64.9000%,35.1000% | |
UN1648 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
Glass Bottle, Gl45 | |
8.3°C |
67.7°C | |
C2H3N | |
0.8561 g/cm3 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
Solvent Blend | |
4 x 2.5 L | |
Acetonitrile 70% and Water 30%, LabReady™ Solvent Blend |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution