missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% formic acid, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS, LabReady™ Solvent Blend
$970.55 - $970.55
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
60-048-215
|
Honeywell-Riedel-de Haen
346684X25L |
2.5 L | Glass Bottle |
Each for $970.55
Case of 4 Each for $3,882.20
|
|
||||
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Chemical Identifiers
75-05-8 | |
41.053 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
Specifications
4.1 | |
75-05-8 | |
0.7767 g/mL at 25°C | |
2.5 L | |
Sweet ether-like | |
Liquid | |
97mmHg | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
LC-MS CHROMASOLV™ |
-46°C | |
Colorless | |
82°C | |
524°C | |
Glass Bottle | |
C2H3N | |
1648 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
99.5 to 100% | |
Formic acid in acetonitrile |
Safety and Handling
Danger