missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Purified Suitable for Histology, Reagents
Acetone, Purified Suitable for Histology, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2102500200E
Specifications
Acetone, Purified Suitable for Histology | |
67-64-1 | |
56°C | |
Steel Drum | |
0.788 | |
MFCD00008765 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
Pure |
-94.7°C | |
Colorless | |
200 L | |
Liquid | |
C3H6O | |
Dimethyl Ketone, 2-Propanone | |
CC(C)=O | |
58.08 |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
Dimethyl Ketone, 2-Propanone | |
CC(C)=O |