missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%
$119.85 - $1002.90
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6000007
|
Honeywell Chemicals
332091LGL |
1 L | Glass Bottle |
Each for $119.85
|
|
||||
6000006
|
Honeywell Chemicals
332091L |
1 L | Plastic Bottle |
Each for $187.63
|
|
||||
6000009
|
Honeywell Chemicals
3320925LGL |
2.5 L | Glass Bottle |
Each for $263.20
|
|
||||
6000008
|
Honeywell Chemicals
3320925L |
2.5 L | Plastic Bottle |
Each for $288.13
|
|
||||
6000011
|
Honeywell Chemicals
332095L |
5 L | Plastic Bottle |
Each for $523.48
|
|
||||
60046219
|
Honeywell Chemicals
332096X1LGL |
6 x 1 L | Glass Bottle |
Case of 6 Each for $618.13
|
|
||||
60046218
|
Honeywell Chemicals
332096X1L |
6 x 1 L | Plastic Bottle |
Case of 6 Each for $990.33
|
|
||||
60046217
|
Honeywell Chemicals
332094X25LGL |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $985.55
|
|
||||
60046216
|
Honeywell Chemicals
332094X25L |
4 x 2.5 L | Plastic Bottle |
Case of 4 Each for $927.30
|
|
||||
6000010
|
Honeywell Chemicals
3320930KG |
30 kg | Plastic Pail |
Each for $482.38
|
|
||||
6000012
|
Honeywell Chemicals
3320960KG |
60 kg | Plastic Pail |
Each for $1,002.90
|
|
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
CH3CO2H | |
UN2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.8% | |
1.049g/mL at 25°C | |
16% at 92°F, 4% at 59°F | |
800°F | |
2.07 (vs air) |
C2H4O2 | |
MFCD00036152 | |
506007 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05g/mol | |
ACS/puriss. p.a. | |
1 L | |
40°C (104°F) | |
Glass Bottle | |
Acetic acid |
Safety and Handling
P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
RTECSNumber : AF1225000