missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.8-100.5%, Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.8-100.5%
$219.60 - $1620.03
Chemical Identifiers
Molecular Formula | C2H4O2 |
---|---|
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | acetasol, acetic acid, glacial, methanecarboxylic acid, vinegar acid, acide acetique, glacial, ethanoic acid, essigsaeure, ethylic acid |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6000031
|
Honeywell-Fluka
272251L |
1 L |
Each for $219.60
|
|
|||||
6000032
|
Honeywell-Fluka
2722525L |
2.5 L |
Each for $371.83
|
|
|||||
60045760
|
Honeywell-Fluka
272256X1L |
6 x 1 L |
Case of 6 Each for $1,012.50
|
|
|||||
6000030
|
Honeywell Chemicals
2722510KG |
10 kg |
Each for $443.05
|
|
|||||
60045759
|
Honeywell-Fluka
272254X25L |
4 x 2.5 L |
Case of 4 Each for $1,241.83
|
|
|||||
6000033
|
Honeywell-Fluka
2722530KG |
30 kg |
Each for $955.60
|
|
|||||
6000035
|
Honeywell-Fluka
2722560KG |
60 kg |
Each for $1,620.03
|
|
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
C2H4O2 | |
MFCD00036152 | |
acetasol, acetic acid, glacial, methanecarboxylic acid, vinegar acid, acide acetique, glacial, ethanoic acid, essigsaeure, ethylic acid | |
CHEBI:15366 | |
CC(O)=O |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
Specifications
64-19-7 | |
CH3CO2H | |
2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
99.8 to 100.5% | |
1.049 g/mL at 25°C | |
16% (92°F), 4% (59°F) | |
800°F | |
2.07 (vs. air) |
C2H4O2 | |
MFCD00036152 | |
506007 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05g/mol | |
puriss. | |
1 L | |
40°C (104°F) | |
Plastic Bottle | |
Acetic Acid, Meets analytical specification of Ph. Eur.BP, USP, FCC |
Safety and Handling
P280-P305 + P351 + P338-P310
H226-H314
EINECSNumber : 200-580-7
RTECSNumber : AF1225000