missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 3% v/v, Reagents
Acetic Acid, 3% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$68.53 - $249.97
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Ethanoic Acid, Ethylic Acid |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-335-916
|
Labchem Inc
CA2035001A |
1 L |
Each for $125.23
|
|
|||||
01-335-915
|
Labchem Inc
CA2035004A |
4 L |
Each for $249.97
|
|
|||||
01-335-914
|
Labchem Inc
CA203500500A |
500 mL |
Each for $68.53
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Ethanoic Acid, Ethylic Acid | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
2.5 | |
100°C | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
3% | |
1 L | |
Liquid | |
Acetic Acid, 3% v/v |
7732-18-5,64-19-7 | |
Colorless | |
Ethanoic Acid, Ethylic Acid | |
O | |
18.02 | |
Reagent | |
Poly Bottle | |
1.0045 |