missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetanilide, BAKER™, J.T. Baker™
A grade of chemicals for laboratory use
Supplier: Avantor J.T.Baker A06906
Specifications
Acetanilide | |
174.00°C | |
C8H9NO | |
250 g | |
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene | |
CC(=O)NC1=CC=CC=C1 | |
135.17 | |
CHEBI:28884 |
103-84-4 | |
Poly Bottle | |
MFCD00008674 | |
1L = 1.22kg | |
FZERHIULMFGESH-UHFFFAOYSA-N | |
N-phenylacetamide | |
904 | |
135.17 |
Chemical Identifiers
103-84-4 | |
135.17 | |
FZERHIULMFGESH-UHFFFAOYSA-N | |
904 | |
N-phenylacetamide |
C8H9NO | |
MFCD00008674 | |
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene | |
CHEBI:28884 | |
CC(=O)NC1=CC=CC=C1 |