missing translation for 'onlineSavingsMsg'
Learn More

Abietic Acid 80.0+%, TCI America™

$91.33 - $643.10

Chemical Identifiers

CAS 514-10-3
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD03423567
InChI Key RSWGJHLUYNHPMX-ONCXSQPRSA-N
Synonym abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
PubChem CID 10569
ChEBI CHEBI:28987
IUPAC Name (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
SMILES CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A000125G
View Documents
TCI America
A000125G
25 g
Each for $91.33
Only null left
 
A0001500G
View Documents
TCI America
A0001500G
500 g
Each for $643.10
Only null left
 
Description

Description

Specifications

Chemical Identifiers

514-10-3
302.46
RSWGJHLUYNHPMX-ONCXSQPRSA-N
10569
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
C20H30O2
MFCD03423567
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
CHEBI:28987
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1

Specifications

164°C
White-Yellow
MFCD03423567
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
302.46
CHEBI:28987
≥80.0% (GC)
Crystalline Powder
514-10-3
C20H30O2
3077
RSWGJHLUYNHPMX-ONCXSQPRSA-N
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
10569
302.46
25 g
Abietic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0675

RTECSNumber : TP8580000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'