Learn More
Applied Biosystems™ ABI LV40™ dCbz Pack DFS Item
Contains 60 dC(bz) ABI LV40™ columns.The dC(bz) ABI LV40™ columns employ the advantages of polystyrene and a low volume column design to deliver high-quality oligonucleotides, while saving time and money.
Supplier:  Applied Biosystems™ 4316342
Catalog No. 4316342
Only null left
Explore available promotions
Description
Description
Save Time and MoneyCompared to conventional columns, the ABI LV40™ columns reduce cycle time by about 10% and the cost of synthesis by about 25%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
  • Proprietary design and manufacturing methods ensure consistency and high quality
  • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
  • Hydrophobic polystryrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis
  • The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length
Specifications
Specifications
Shipped at Room Temperature
60 Columns
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.