Learn More
Electron Microscopy Round Cover Slip German Glass #1.5, 8 mm
Supplier: Electron Microscopy Sciences 7229608
These coverglass which are made from German glass can be used for growing and culturing cells that normally have poor adhesion to plastic or regular glass surfaces. Available with #1.5 (0.16 0.19 mm) thickness. 8 mm fits inside the 24 well plates.
Diameter: 8 mm