missing translation for 'onlineSavingsMsg'
Learn More

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%

CAS: 54745-74-3 | C6H12ClNO | 149.62 g/mol

Supplier:  Thermo Scientific Chemicals H6382506

 View more versions of this product

Catalog No. AAH6382506


Only null left
Explore available promotions

Description

Description

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride
C6H12ClNO
5 g
XADOTNAXKKFKDY-UHFFFAOYNA-N
8-oxa-3-azabicyclo[3.2.1]octane hydrochloride
21983536
97%
54745-74-3
MFCD17926456,MFCD09800611
8-oxa-3-azabicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclo 3,2,1 octane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane hcl, 8-oxa-3-azabicyclo 3.2.1 octanehydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane, hydrochloride, 8-oxa-3-aza-bicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclooctane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane-hcl, 8-oxa-3-aza-bicyclo 3.2.1 octane hcl
Cl.C1CC2CNCC1O2
149.62
149.62

Chemical Identifiers

54745-74-3
149.62
XADOTNAXKKFKDY-UHFFFAOYNA-N
21983536
C6H12ClNO
MFCD17926456,MFCD09800611
8-oxa-3-azabicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclo 3,2,1 octane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane hcl, 8-oxa-3-azabicyclo 3.2.1 octanehydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane, hydrochloride, 8-oxa-3-aza-bicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclooctane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane-hcl, 8-oxa-3-aza-bicyclo 3.2.1 octane hcl
Cl.C1CC2CNCC1O2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only