missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Azahypoxanthine, 98%
CAS: 2683-90-1 | C4H3N5O | 137.10 g/mol
$335.12 - $1308.25
Chemical Identifiers
CAS | 2683-90-1 |
---|---|
Molecular Formula | C4H3N5O |
Molecular Weight (g/mol) | 137.10 |
MDL Number | MFCD00005804 |
InChI Key | OEEYCNOOAHGFHL-UHFFFAOYSA-N |
Synonym | 8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine |
PubChem CID | 75895 |
IUPAC Name | 2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one |
SMILES | O=C1N=CN=C2NNN=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC202590010
|
Thermo Scientific Chemicals
202590010 |
1 g |
Each for $335.12
|
|
|||||
AC202590050
|
Thermo Scientific Chemicals
202590050 |
5 g |
Each for $1,308.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2683-90-1 | |
137.10 | |
OEEYCNOOAHGFHL-UHFFFAOYSA-N | |
75895 | |
O=C1N=CN=C2NNN=C12 |
C4H3N5O | |
MFCD00005804 | |
8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine | |
2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one |
Specifications
2683-90-1 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00005804 | |
26, IV, 4130 | |
Solubility in water: insoluble. Other solubilities: soluble in alkaline solution at ph >9 | |
O=C1N=CN=C2NNN=C12 | |
137.10 | |
137.1 | |
Fine Crystalline Powder |
97.5 | |
Beige to Yellow | |
97.5% min. (HPLC) | |
C4H3N5O | |
1 g | |
8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine | |
OEEYCNOOAHGFHL-UHFFFAOYSA-N | |
2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one | |
75895 | |
98% | |
8-Azahypoxanthine, 98% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 220-244-3
TSCA : TSCA
RUO – Research Use Only