missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-5-methylbenzo[b]furan, 97%

CAS: 35700-48-2 | C9H7BrO | 211.058 g/mol

$180.05 - $229.49

Chemical Identifiers

CAS 35700-48-2
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD11877828
InChI Key TZUMDTURHSPEAY-UHFFFAOYSA-N
Synonym 7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
PubChem CID 21071801
IUPAC Name 7-bromo-5-methyl-1-benzofuran
SMILES CC1=CC(=C2C(=C1)C=CO2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH34347MD
View Documents
Thermo Scientific Chemicals
H34347MD
250 mg
Each for $180.05
Only null left
 
AAH3434703
View Documents
Thermo Scientific Chemicals
H3434703
1 g
Each for $229.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35700-48-2
211.058
TZUMDTURHSPEAY-UHFFFAOYSA-N
21071801
CC1=CC(=C2C(=C1)C=CO2)Br
C9H7BrO
MFCD11877828
7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
7-bromo-5-methyl-1-benzofuran

Specifications

35700-48-2
MFCD11877828
7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
CC1=CC(=C2C(=C1)C=CO2)Br
211.058
211.06
7-Bromo-5-methylbenzo[b]furan
C9H7BrO
250 mg
TZUMDTURHSPEAY-UHFFFAOYSA-N
7-bromo-5-methyl-1-benzofuran
21071801
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only