missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Benzyloxy-6-methoxy-4(3H)-quinazolinone, 96%
CAS: 179688-01-8 | C16H14N2O3 | 282.299 g/mol
$87.22 - $291.27
Chemical Identifiers
CAS | 179688-01-8 |
---|---|
Molecular Formula | C16H14N2O3 |
Molecular Weight (g/mol) | 282.299 |
MDL Number | MFCD04115119 |
InChI Key | ZCUFFSHMOAEEIL-UHFFFAOYSA-N |
Synonym | 7-benzyloxy-6-methoxyquinazolin-4 3h-one, 6-methoxy-7-benzyloxyquinazolin-4-one, 7-benzyloxy-6-methoxy-3h-quinazolin-4-one, 7-benzyloxy-6-methoxyquinazolin-4-ol, 7-benzyloxy-6-methoxyquinazolin-4 1h-one, 4 3h-quinazolinone, 6-methoxy-7-phenylmethoxy, 7-benzyloxy-6-methoxy-4 3h-quinazolinone, 4 1h-quinazolinone, 6-methoxy-7-phenylmethoxy |
PubChem CID | 10540849 |
IUPAC Name | 6-methoxy-7-phenylmethoxy-1H-quinazolin-4-one |
SMILES | COC1=C(C=C2C(=C1)C(=O)N=CN2)OCC3=CC=CC=C3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3413103
|
Thermo Scientific Chemicals
H3413103 |
1 g |
Each for $87.22
|
|
|||||
AAH3413106
|
Thermo Scientific Chemicals
H3413106 |
5 g |
Each for $291.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
179688-01-8 | |
282.299 | |
ZCUFFSHMOAEEIL-UHFFFAOYSA-N | |
10540849 | |
COC1=C(C=C2C(=C1)C(=O)N=CN2)OCC3=CC=CC=C3 |
C16H14N2O3 | |
MFCD04115119 | |
7-benzyloxy-6-methoxyquinazolin-4 3h-one, 6-methoxy-7-benzyloxyquinazolin-4-one, 7-benzyloxy-6-methoxy-3h-quinazolin-4-one, 7-benzyloxy-6-methoxyquinazolin-4-ol, 7-benzyloxy-6-methoxyquinazolin-4 1h-one, 4 3h-quinazolinone, 6-methoxy-7-phenylmethoxy, 7-benzyloxy-6-methoxy-4 3h-quinazolinone, 4 1h-quinazolinone, 6-methoxy-7-phenylmethoxy | |
6-methoxy-7-phenylmethoxy-1H-quinazolin-4-one |
Specifications
179688-01-8 | |
MFCD04115119 | |
7-benzyloxy-6-methoxyquinazolin-4 3h-one, 6-methoxy-7-benzyloxyquinazolin-4-one, 7-benzyloxy-6-methoxy-3h-quinazolin-4-one, 7-benzyloxy-6-methoxyquinazolin-4-ol, 7-benzyloxy-6-methoxyquinazolin-4 1h-one, 4 3h-quinazolinone, 6-methoxy-7-phenylmethoxy, 7-benzyloxy-6-methoxy-4 3h-quinazolinone, 4 1h-quinazolinone, 6-methoxy-7-phenylmethoxy | |
COC1=C(C=C2C(=C1)C(=O)N=CN2)OCC3=CC=CC=C3 | |
282.299 | |
284.31 | |
7-Benzyloxy-6-methoxy-4(3H)-quinazolinone |
C16H14N2O3 | |
1 g | |
ZCUFFSHMOAEEIL-UHFFFAOYSA-N | |
6-methoxy-7-phenylmethoxy-1H-quinazolin-4-one | |
10540849 | |
96% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only