missing translation for 'onlineSavingsMsg'
Learn More

7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™

$222.42 - $222.42

Chemical Identifiers

CAS 156311-85-2
Molecular Formula C11H21F6N7OP2
Molecular Weight (g/mol) 443.275
MDL Number MFCD09263289
InChI Key RQBNNDQCKMIUQJ-UHFFFAOYSA-N
Synonym 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+%
PubChem CID 21894514
IUPAC Name tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
SMILES CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2660906
View Documents
Thermo Scientific Chemicals
H2660906
5 g
Each for $222.42
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

156311-85-2
443.275
RQBNNDQCKMIUQJ-UHFFFAOYSA-N
21894514
CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F
C11H21F6N7OP2
MFCD09263289
3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+%
tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate

Specifications

156311-85-2
C11H21F6N7OP2
5 g
RQBNNDQCKMIUQJ-UHFFFAOYSA-N
tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
21894514
≥98%
∽180°C (decomposition)
MFCD09263289
3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+%
CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F
443.275
443.27
7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)Phosphonium Hexafluorophosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'