missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%

CAS: 28226-22-4 | C8H8N2O3 | 180.163 g/mol

$62.51 - $162.77

Chemical Identifiers

CAS 28226-22-4
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD07698591
InChI Key GZAJZBARYACGSO-UHFFFAOYSA-N
Synonym 6-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-6-nitro, 2h-1,4-benzoxazine, 3,4-dihydro-6-nitro, 6-nitro-2,3-dihydrobenz-1,4-oxazine, 6-nitro-2h,3h,4h-benzo e 1,4-oxazine, 6-nitro-3,4-dihydro-2h-1,4 benzoxazine, 6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 3,4-dihydro-6-nitro-2h-benzo b 1,4 oxazine, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronicacid,pinacolester
PubChem CID 7062225
IUPAC Name 6-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=C(N1)C=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH33900MD
View Documents
Thermo Scientific Chemicals
H33900MD
250 mg
Each for $62.51
Only null left
 
AAH3390003
View Documents
Thermo Scientific Chemicals
H3390003
1 g
Each for $162.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28226-22-4
180.163
GZAJZBARYACGSO-UHFFFAOYSA-N
7062225
C1COC2=C(N1)C=C(C=C2)[N+](=O)[O-]
C8H8N2O3
MFCD07698591
6-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-6-nitro, 2h-1,4-benzoxazine, 3,4-dihydro-6-nitro, 6-nitro-2,3-dihydrobenz-1,4-oxazine, 6-nitro-2h,3h,4h-benzo e 1,4-oxazine, 6-nitro-3,4-dihydro-2h-1,4 benzoxazine, 6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 3,4-dihydro-6-nitro-2h-benzo b 1,4 oxazine, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronicacid,pinacolester
6-nitro-3,4-dihydro-2H-1,4-benzoxazine

Specifications

28226-22-4
C8H8N2O3
250 mg
GZAJZBARYACGSO-UHFFFAOYSA-N
6-nitro-3,4-dihydro-2H-1,4-benzoxazine
7062225
97%
117°C to 119°C
MFCD07698591
6-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-6-nitro, 2h-1,4-benzoxazine, 3,4-dihydro-6-nitro, 6-nitro-2,3-dihydrobenz-1,4-oxazine, 6-nitro-2h,3h,4h-benzo e 1,4-oxazine, 6-nitro-3,4-dihydro-2h-1,4 benzoxazine, 6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 3,4-dihydro-6-nitro-2h-benzo b 1,4 oxazine, 1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronicacid,pinacolester
C1COC2=C(N1)C=C(C=C2)[N+](=O)[O-]
180.163
180.2
6-Nitro-3,4-dihydro-2H-1,4-benzoxazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only