missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Sulfosalicylic Acid, Dihydrate, Crystal, ACS, 99.0-101.0%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S171025KG
Specifications
5965-83-3 | |
0.001 | |
C9H14O6S | |
NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
2-hydroxy-5-sulfobenzoic acid; bis(methane) | |
99 to 101% | |
0.0002 |
1 | |
Amber Glass Bottle | |
2.5 kg | |
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O | |
250.27 | |
ACS | |
Crystalline Powder or Lumps |
Chemical Identifiers
5965-83-3 | |
250.27 | |
2-hydroxy-5-sulfobenzoic acid; bis(methane) |
C9H14O6S | |
NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.