missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzothiazole, 96%, Thermo Scientific Chemicals

$117.22 - $117.22

Chemical Identifiers

CAS 2942-07-6
MDL Number MFCD00085891
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3359403
View Documents
Thermo Scientific Chemicals
H3359403
1 g
Each for $117.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2942-07-6
MFCD00085891

Specifications

2942-07-6
C12H18ClNO2
1 g
QVFHQENRNSAHEK-UHFFFAOYSA-M
2-benzoyloxyethyl(trimethyl)azanium;chloride
76298
96%
157°C to 162°C
MFCD00085891
benzoylcholine chloride, 2-hydroxyethyl trimethylammonium chloride benzoate, choline, chloride, benzoate, 2-benzoyloxy-n,n,n-trimethylethanaminium chloride, 2-benzoyloxyethyltrimethylammonium chloride, 2-benzoyloxy ethyl trimethylazanium chloride, 2-benzyloxy-n,n,n-trimethylethanaminium chloride, 2-benzyloxy-n,n,n-trimethylethanaminimum chloride, ammonium, 2-hydroxyethyl trimethyl-, chloride, benzoate, 2-trimethylamino ethyl benzoate, chloride
C[N+](C)(C)CCOC(=O)C1=CC=CC=C1.[Cl-]
243.73
180.18
5-Nitrobenzothiazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only