missing translation for 'onlineSavingsMsg'
Learn More

5-n-Octyl-2,2'-bithiophene 97.0+%, TCI America™

$305.70 - $1004.73

Chemical Identifiers

CAS 93164-73-9
Molecular Formula C16H22S2
Molecular Weight (g/mol) 278.472
InChI Key ZYEIXHZLTHTEOI-UHFFFAOYSA-N
PubChem CID 56973709
IUPAC Name 2-octyl-5-thiophen-2-ylthiophene
SMILES CCCCCCCCC1=CC=C(S1)C2=CC=CS2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O03831G
View Documents
TCI America
O03831G
1 g
Each for $305.70
Only null left
 
O03835G
View Documents
TCI America
O03835G
5 g
Each for $1,004.73
Only null left
 
Description

Description

Specifications

Chemical Identifiers

93164-73-9
278.472
56973709
CCCCCCCCC1=CC=C(S1)C2=CC=CS2
C16H22S2
ZYEIXHZLTHTEOI-UHFFFAOYSA-N
2-octyl-5-thiophen-2-ylthiophene

Specifications

93164-73-9
Yellow
1 g
CCCCCCCCC1=CC=C(S1)C2=CC=CS2
278.472
278.47
Crystalline Lumps
29°C
C16H22S2
ZYEIXHZLTHTEOI-UHFFFAOYSA-N
2-octyl-5-thiophen-2-ylthiophene
56973709
≥97.0% (GC)
5-n-Octyl-2,2′-bithiophene
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'