Learn More
5-Methylindole, 99%
CAS: 614-96-0 | C9H9N | 131.178 g/mol
$120.21 - $439.56
Chemical Identifiers
CAS | 614-96-0 |
---|---|
Molecular Formula | C9H9N |
Molecular Weight (g/mol) | 131.178 |
MDL Number | MFCD00005680 |
InChI Key | YPKBCLZFIYBSHK-UHFFFAOYSA-N |
Synonym | 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238 |
PubChem CID | 11978 |
IUPAC Name | 5-methyl-1H-indole |
SMILES | CC1=CC2=C(C=C1)NC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0689303
|
Thermo Scientific Chemicals
L0689303 |
1 g |
N/A
|
|
|||||
AAL0689306
|
Thermo Scientific Chemicals
L0689306 |
5 g |
N/A
|
|
|||||
Description
5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents and heat.
Chemical Identifiers
614-96-0 | |
131.178 | |
YPKBCLZFIYBSHK-UHFFFAOYSA-N | |
11978 | |
CC1=CC2=C(C=C1)NC=C2 |
C9H9N | |
MFCD00005680 | |
5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238 | |
5-methyl-1H-indole |
Specifications
614-96-0 | |
100°C (3 mmHg) | |
MFCD00005680 | |
2228 | |
5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238 | |
YPKBCLZFIYBSHK-UHFFFAOYSA-N | |
5-methyl-1H-indole | |
11978 | |
99% |
58°C to 61°C | |
C9H9N | |
1 g | |
Light sensitive | |
Insoluble in water. | |
CC1=CC2=C(C=C1)NC=C2 | |
131.178 | |
131.18 | |
5-Methylindole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 210-400-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only