missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+%

CAS: 848613-91-2 | C16H31NSSn | 388.201 g/mol

$182.59 - $627.52

Chemical Identifiers

CAS 848613-91-2
Molecular Formula C16H31NSSn
Molecular Weight (g/mol) 388.201
MDL Number MFCD09025809
InChI Key LALGELPHLJBAEK-UHFFFAOYSA-N
Synonym 5-methyl-2-tributylstannyl thiazole, 5-methyl-2-tributylstannyl-1,3-thiazole, 5-methyl-2-tri-n-butylstannyl thiazole, acmc-209px3, 5-methyl-2-tributylstannyl-thiazole, thiazole, 5-methyl-2-tributylstannyl, 5-methyl-2-tributylstannanyl-thiazole, 5-methyl-2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl-5-methylthiazole
PubChem CID 16427131
IUPAC Name tributyl-(5-methyl-1,3-thiazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=C(S1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51531MD
View Documents
Thermo Scientific Chemicals
H51531MD
250 mg
Each for $182.59
Only null left
 
AAH5153103
View Documents
Thermo Scientific Chemicals
H5153103
1 g
Each for $627.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

848613-91-2
388.201
LALGELPHLJBAEK-UHFFFAOYSA-N
16427131
CCCC[Sn](CCCC)(CCCC)C1=NC=C(S1)C
C16H31NSSn
MFCD09025809
5-methyl-2-tributylstannyl thiazole, 5-methyl-2-tributylstannyl-1,3-thiazole, 5-methyl-2-tri-n-butylstannyl thiazole, acmc-209px3, 5-methyl-2-tributylstannyl-thiazole, thiazole, 5-methyl-2-tributylstannyl, 5-methyl-2-tributylstannanyl-thiazole, 5-methyl-2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl-5-methylthiazole
tributyl-(5-methyl-1,3-thiazol-2-yl)stannane

Specifications

848613-91-2
MFCD09025809
UN2788
LALGELPHLJBAEK-UHFFFAOYSA-N
tributyl-(5-methyl-1,3-thiazol-2-yl)stannane
16427131
≥90%
C16H31NSSn
250 mg
5-methyl-2-tributylstannyl thiazole, 5-methyl-2-tributylstannyl-1,3-thiazole, 5-methyl-2-tri-n-butylstannyl thiazole, acmc-209px3, 5-methyl-2-tributylstannyl-thiazole, thiazole, 5-methyl-2-tributylstannyl, 5-methyl-2-tributylstannanyl-thiazole, 5-methyl-2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl-5-methylthiazole
CCCC[Sn](CCCC)(CCCC)C1=NC=C(S1)C
388.201
388.19
5-Methyl-2-(tri-n-butylstannyl)thiazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only