missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxynorbornane 2,6-Lactone 98.0+%, TCI America™
$54.06 - $145.40
Chemical Identifiers
CAS | 92343-46-9 |
---|---|
Molecular Formula | C8H10O3 |
Molecular Weight (g/mol) | 154.165 |
MDL Number | MFCD00988184 |
InChI Key | SCDJKGVPVLZLED-UHFFFAOYSA-N |
Synonym | 6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one |
PubChem CID | 432639 |
SMILES | C1C2CC3C1C(C2O)OC3=O |
Chemical Identifiers
92343-46-9 | |
154.165 | |
SCDJKGVPVLZLED-UHFFFAOYSA-N | |
432639 |
C8H10O3 | |
MFCD00988184 | |
6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one | |
C1C2CC3C1C(C2O)OC3=O |
Specifications
92343-46-9 | |
Yellow | |
MFCD00988184 | |
6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one | |
C1C2CC3C1C(C2O)OC3=O | |
432639 | |
≥98.0% (GC) | |
5-Hydroxynorbornane 2,6-Lactone |
158°C | |
C8H10O3 | |
5 g | |
SCDJKGVPVLZLED-UHFFFAOYSA-N | |
154.165 | |
154.17 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6966
TSCA : No