missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Bromo-m-terphenyl 98.0+%, TCI America™
$153.06 - $461.10
Chemical Identifiers
CAS | 103068-20-8 |
---|---|
Molecular Formula | C18H13Br |
Molecular Weight (g/mol) | 309.21 |
MDL Number | MFCD00196170 |
InChI Key | IOPQERQQZZREDR-UHFFFAOYSA-N |
PubChem CID | 10070257 |
IUPAC Name | 3-bromo-5-phenyl-1,1'-biphenyl |
SMILES | BrC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
103068-20-8 | |
309.21 | |
IOPQERQQZZREDR-UHFFFAOYSA-N | |
3-bromo-5-phenyl-1,1'-biphenyl |
C18H13Br | |
MFCD00196170 | |
10070257 | |
BrC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
103068-20-8 | |
White | |
MFCD00196170 | |
3152 | |
IOPQERQQZZREDR-UHFFFAOYSA-N | |
3-bromo-5-phenyl-1,1'-biphenyl | |
10070257 | |
≥98.0% (GC) | |
5′-Bromo-m-terphenyl |
111°C | |
C18H13Br | |
5 g | |
1-Bromo-3,5-diphenylbenzene, 5′C-Bromo-1,1′C:3′C,1′C′C-terphenyl | |
BrC1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
309.21 | |
309.21 | |
Crystalline Powder |
Safety and Handling
TSCA : No