missing translation for 'onlineSavingsMsg'
Learn More

5″-Bromo-2,2':5',2″-terthiophene-5-carboxaldehyde 94.0+%, TCI America™

$480.92 - $1916.25

Chemical Identifiers

CAS 161726-69-8
Molecular Formula C13H7BrOS3
Molecular Weight (g/mol) 355.28
MDL Number MFCD02093442
InChI Key DSYKMLTVEGWLFR-UHFFFAOYSA-N
PubChem CID 14942412
IUPAC Name 5-{5'-bromo-[2,2'-bithiophen]-5-yl}thiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20581G
View Documents
TCI America
B20581G
1 g
Each for $480.92
Only null left
 
B20585G
View Documents
TCI America
B20585G
5 g
Each for $1,916.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

161726-69-8
355.28
DSYKMLTVEGWLFR-UHFFFAOYSA-N
5-{5'-bromo-[2,2'-bithiophen]-5-yl}thiophene-2-carbaldehyde
C13H7BrOS3
MFCD02093442
14942412
BrC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C=O

Specifications

161726-69-8
C13H7BrOS3
1 g
DSYKMLTVEGWLFR-UHFFFAOYSA-N
5-{5'-bromo-[2,2'-bithiophen]-5-yl}thiophene-2-carbaldehyde
14942412
≥94.0% (GC)
5′′-Bromo-2,2′:5′,2′′-terthiophene-5-carboxaldehyde
164°C
MFCD02093442
5-Bromo-5′C′C-formyl-2,2′C:5′C,2′C′C-terthiophene
BrC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C=O
355.28
355.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'