missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-(4-aminophenyl)benzimidazole 98.0+%, TCI America™
$83.14 - $338.27
Chemical Identifiers
CAS | 7621-86-5 |
---|---|
Molecular Formula | C13H12N4 |
Molecular Weight (g/mol) | 224.27 |
MDL Number | MFCD00043996,MFCD00451475 |
InChI Key | XAFOTXWPFVZQAZ-UHFFFAOYSA-N |
PubChem CID | 24260 |
IUPAC Name | 2-(4-aminophenyl)-1H-1,3-benzodiazol-6-amine |
SMILES | NC1=CC=C(C=C1)C1=NC2=CC=C(N)C=C2N1 |
Chemical Identifiers
7621-86-5 | |
224.27 | |
XAFOTXWPFVZQAZ-UHFFFAOYSA-N | |
2-(4-aminophenyl)-1H-1,3-benzodiazol-6-amine |
C13H12N4 | |
MFCD00043996,MFCD00451475 | |
24260 | |
NC1=CC=C(C=C1)C1=NC2=CC=C(N)C=C2N1 |
Specifications
7621-86-5 | |
White-Yellow | |
MFCD00043996,MFCD00451475 | |
XAFOTXWPFVZQAZ-UHFFFAOYSA-N | |
2-(4-aminophenyl)-1H-1,3-benzodiazol-6-amine | |
24260 | |
≥98.0% (HPLC) | |
5-Amino-2-(4-aminophenyl)benzimidazole |
236°C | |
C13H12N4 | |
1 g | |
NC1=CC=C(C=C1)C1=NC2=CC=C(N)C=C2N1 | |
224.27 | |
224.27 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : DD5421200
TSCA : Yes