missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1,2,3-thiadiazole 98.0+%, TCI America™

$108.11 - $322.48

Chemical Identifiers

CAS 4100-41-8
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.127
MDL Number MFCD00082810
InChI Key PVGHNTXQMCYYGF-UHFFFAOYSA-N
PubChem CID 77736
IUPAC Name thiadiazol-5-amine
SMILES C1=C(SN=N1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A23545G
View Documents
TCI America
A23545G
5 g
Each for $108.11
Only null left
 
A235425G
View Documents
TCI America
A235425G
25 g
Each for $322.48
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4100-41-8
101.127
PVGHNTXQMCYYGF-UHFFFAOYSA-N
thiadiazol-5-amine
C2H3N3S
MFCD00082810
77736
C1=C(SN=N1)N

Specifications

4100-41-8
C2H3N3S
5 g
C1=C(SN=N1)N
101.127
101.13
Crystalline Powder
142°C
MFCD00082810
PVGHNTXQMCYYGF-UHFFFAOYSA-N
thiadiazol-5-amine
77736
≥98.0% (HPLC)
5-Amino-1,2,3-thiadiazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'