missing translation for 'onlineSavingsMsg'
Learn More

5,5″-Dibromo-2,2':5',2″-terthiophene 97.0+%, TCI America™

$311.81 - $1069.60

Chemical Identifiers

CAS 98057-08-0
Molecular Formula C12H6Br2S3
Molecular Weight (g/mol) 406.168
MDL Number MFCD00219109
InChI Key KXFPYYJGYVYXIB-UHFFFAOYSA-N
Synonym 2,5-Bis(5-bromo-2-thienyl)thiophene
PubChem CID 176690
IUPAC Name 2,5-bis(5-bromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1)C2=CC=C(S2)Br)C3=CC=C(S3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D40501G
View Documents
TCI America
D40501G
1 g
Each for $311.81
Only null left
 
D40505G
View Documents
TCI America
D40505G
5 g
Each for $1,069.60
Only null left
 
Description

Description

Specifications

Chemical Identifiers

98057-08-0
406.168
KXFPYYJGYVYXIB-UHFFFAOYSA-N
176690
C1=C(SC(=C1)C2=CC=C(S2)Br)C3=CC=C(S3)Br
C12H6Br2S3
MFCD00219109
2,5-Bis(5-bromo-2-thienyl)thiophene
2,5-bis(5-bromothiophen-2-yl)thiophene

Specifications

98057-08-0
Yellow
MFCD00219109
2,5-Bis(5-bromo-2-thienyl)thiophene
C1=C(SC(=C1)C2=CC=C(S2)Br)C3=CC=C(S3)Br
406.168
406.17
Crystalline Powder
158°C
C12H6Br2S3
1 g
KXFPYYJGYVYXIB-UHFFFAOYSA-N
2,5-bis(5-bromothiophen-2-yl)thiophene
176690
≥97.0% (GC)
5,5′′-Dibromo-2,2′:5′,2′′-terthiophene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents