missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin 90.0+%, TCI America™

$259.11 - $1502.43

Chemical Identifiers

CAS 119730-06-2
Molecular Formula C46H32N4O2
Molecular Weight (g/mol) 672.788
MDL Number MFCD02093480
InChI Key JYTWPMXQCKOXJN-UHFFFAOYSA-N
Synonym 5-(4-Carbomethoxyphenyl)-10,15,20-triphenylporphyrin, 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenyl-21H,23H-porphine
PubChem CID 11181451
IUPAC Name methyl 4-(10,15,20-triphenyl-23,24-dihydroporphyrin-5-yl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C2=C3C=CC(=N3)C(=C4C=CC(=C(C5=CC=C(N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)N4)C9=CC=CC=C9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1338100MG
View Documents
TCI America
M1338100MG
100 mg
Each for $259.11
Only null left
 
M13381G
View Documents
TCI America
M13381G
1 g
Each for $1,502.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

119730-06-2
672.788
JYTWPMXQCKOXJN-UHFFFAOYSA-N
11181451
COC(=O)C1=CC=C(C=C1)C2=C3C=CC(=N3)C(=C4C=CC(=C(C5=CC=C(N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)N4)C9=CC=CC=C9
C46H32N4O2
MFCD02093480
5-(4-Carbomethoxyphenyl)-10,15,20-triphenylporphyrin, 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenyl-21H,23H-porphine
methyl 4-(10,15,20-triphenyl-23,24-dihydroporphyrin-5-yl)benzoate

Specifications

119730-06-2
C46H32N4O2
100 mg
JYTWPMXQCKOXJN-UHFFFAOYSA-N
methyl 4-(10,15,20-triphenyl-23,24-dihydroporphyrin-5-yl)benzoate
11181451
≥90.0% (HPLC)
5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin
Blue-Purple
MFCD02093480
5-(4-Carbomethoxyphenyl)-10,15,20-triphenylporphyrin, 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenyl-21H,23H-porphine
COC(=O)C1=CC=C(C=C1)C2=C3C=CC(=N3)C(=C4C=CC(=C(C5=CC=C(N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)N4)C9=CC=CC=C9
672.788
672.79
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'