missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ BRAND™ BRANDplates™ Multiwell Plates for Cell Culture Inserts, - 6-well Specialized Plate
Sterile and individually wrapped and available with either untreated or cell culture treated surface.
$828.09 - $871.91
Specifications
Closure Included | Yes |
---|---|
Bottom Shape | Flat |
Capacity (Metric) | 10 mL |
Color | Transparent |
Culture Area | 855 mm2 |
Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity | |||||
14-380-866
|
BrandTech™ BRAND™
782881 |
pureGrade S |
Pack of 10 Each for $828.09
|
|
|||||
14-380-868
|
BrandTech™ BRAND™
782891 |
cellGrade plus |
Pack of 10 Each for $871.91
|
|
|||||
Description
- BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
- Available with pureGrade™ S and cellGrade™ plus surfaces
- 6-well special plate design connects 4 wells into one large, elongated well, allowing simultaneous medium supply to all 4 inserts in a strip
- Ideal for insert strips with inlet channels, it also supports single inserts and combinations of 2 or 3 connected inserts
- Guide ridges for perfect insert fit
- Designated feeding ports for easy media changes, suitable for robotics/automation
- 10 plates per pack (individually wrapped with lid)
Surface Information:
- pureGrade™ S: Non-treated, hydrophobic surface with medium binding capacity (ELISA). Especially suited for bacteriological assays.
- cellGrade™ plus: Specifically designed for the cultivation of fastidious cell lines. Features carboxyl, hydroxy, and free amino groups on the surface, offering a protein-like composition that facilitates direct cell attachment and spreading.
Applications:
- Ideal for 3D tissue cultures
Compliance
ANSI/SLAS standards 1 and 4
Specifications
Yes | |
10 mL | |
855 mm2 | |
Sterile | |
6 | |
Insert system |
Flat | |
Transparent | |
Polystyrene | |
Cell Culture Insert System | |
3D tissue culture | |
19.5 mm |