Learn More
National Safety Apparel™ 40 Cal ArcGuard™ RevoLite™ Bib Overalls
RevoLite; bib overalls are 50% lighter than standard 40 cal/cm2 Arc Flash PPE
$1516.61 - $2018.63
Specifications
Closure Type FR Hook and Loop with Buckle
Color Olive
Material RevoLite™
Seam Type Stitched
Suspender Adjustable
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17001325
View Documents
National Safety Apparel™
C45NPQHLTSM32
Small
Each for $1,516.61
Only null left
17001326
View Documents
National Safety Apparel™
C45NPQHLTMD32
Medium
Each for $1,516.61
Only null left
17001327
View Documents
National Safety Apparel™
C45NPQHLTLG32
Large
Each for $1,516.61
Only null left
17001328
View Documents
National Safety Apparel™
C45NPQHLTXL32
X-Large
Each for $1,516.61
Only null left
17001329
View Documents
National Safety Apparel™
C45NPQHLT2X32
2X-Large
Each for $1,668.28
Only null left
17001330
View Documents
National Safety Apparel™
C45NPQHLT3X32
3X-Large
Each for $1,668.28
Only null left
17001331
View Documents
National Safety Apparel™
C45NPQHLT4X32
4X-Large
Each for $1,835.12
Only null left
17001332
View Documents
National Safety Apparel™
C45NPQHLT5X32
5X-Large
Each for $2,018.63
Only null left
Description
Description
  • Inherently flame resistant for FR protection
  • FR fabric will not melt, drip, or ignite
  • Quick disconnect and easy adjustable shoulder straps
  • Heavy duty 26 in. zipper closure on each leg
  • Expandable leg opening to fit over boots
  • Garments embroidered with arc rating
  • Available in a convenient kit
Specifications
Specifications
FR Hook and Loop with Buckle
RevoLite™
Adjustable
Bib Overalls
Flame
Reusable
Olive
Stitched
40 cal/cm²
Standard
CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'