missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylsulfonyl)phenylacetic acid, 98%

CAS: 1099597-82-6 | C9H7F3O4S | 268.206 g/mol

$220.66 - $651.40

Chemical Identifiers

CAS 1099597-82-6
Molecular Formula C9H7F3O4S
Molecular Weight (g/mol) 268.206
MDL Number MFCD04973012
InChI Key FIEUCHICQSJBAU-UHFFFAOYSA-N
Synonym 4-trifluoromethylsulfony phenylacetic acid, 2-4-trifluoromethyl sulfonyl phenyl acetic acid, 4-trifluoromethanesulfonylphenyl acetic acid, 4-trifluoromethylsulfonyl phenylacetic acid, 4-trifluoromethanesulfonyl phenyl acetic acid
PubChem CID 40427164
IUPAC Name 2-[4-(trifluoromethylsulfonyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)S(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3229503
View Documents
Thermo Scientific Chemicals
H3229503
1 g
Each for $220.66
Only null left
 
AAH3229506
View Documents
Thermo Scientific Chemicals
H3229506
5 g
Each for $651.40
Only null left
 
Description

Description

4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethylsulfonyl)phenylacetic acid, is used as a primary and secondary intermediate.

Solubility
Slightly soluble in water, soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

1099597-82-6
268.206
FIEUCHICQSJBAU-UHFFFAOYSA-N
40427164
C1=CC(=CC=C1CC(=O)O)S(=O)(=O)C(F)(F)F
C9H7F3O4S
MFCD04973012
4-trifluoromethylsulfony phenylacetic acid, 2-4-trifluoromethyl sulfonyl phenyl acetic acid, 4-trifluoromethanesulfonylphenyl acetic acid, 4-trifluoromethylsulfonyl phenylacetic acid, 4-trifluoromethanesulfonyl phenyl acetic acid
2-[4-(trifluoromethylsulfonyl)phenyl]acetic acid

Specifications

1099597-82-6
C9H7F3O4S
1 g
Slightly soluble in water,soluble in organic solvents.
C1=CC(=CC=C1CC(=O)O)S(=O)(=O)C(F)(F)F
268.206
268.21
4-(Trifluoromethylsulfonyl)phenylacetic acid
125°C to 128°C
MFCD04973012
4-trifluoromethylsulfony phenylacetic acid, 2-4-trifluoromethyl sulfonyl phenyl acetic acid, 4-trifluoromethanesulfonylphenyl acetic acid, 4-trifluoromethylsulfonyl phenylacetic acid, 4-trifluoromethanesulfonyl phenyl acetic acid
FIEUCHICQSJBAU-UHFFFAOYSA-N
2-[4-(trifluoromethylsulfonyl)phenyl]acetic acid
40427164
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only