missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfonamidophenylhydrazine hydrochloride, 97%

CAS: 17852-52-7 | C6H10ClN3O2S | 223.675 g/mol

Supplier:  Thermo Scientific Chemicals B2001509

 View more versions of this product

Catalog No. AAB2001509


Only null left
Explore available promotions

Description

Description

It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

4-Sulfonamidophenylhydrazine hydrochloride
217°C to 219°C (decomposition)
MFCD00052262
Hygroscopic
Soluble in water.
C1=CC(=CC=C1NN)S(=O)(=O)N.Cl
223.675
223.68
17852-52-7
C6H10ClN3O2S
10 g
4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride
IKEURONJLPUALY-UHFFFAOYSA-N
4-hydrazinylbenzenesulfonamide;hydrochloride
2794567
97%

Chemical Identifiers

17852-52-7
223.675
IKEURONJLPUALY-UHFFFAOYSA-N
2794567
C1=CC(=CC=C1NN)S(=O)(=O)N.Cl
C6H10ClN3O2S
MFCD00052262
4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride
4-hydrazinylbenzenesulfonamide;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only