missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)phenol, 97%

CAS: 774-94-7 | C6H5F5OS | 220.16 g/mol

$201.25 - $792.62

Chemical Identifiers

CAS 774-94-7
Molecular Formula C6H5F5OS
Molecular Weight (g/mol) 220.16
MDL Number MFCD03788516
InChI Key XHJLGVIUMCBMHL-UHFFFAOYSA-N
Synonym 4-pentafluorothio phenol, 4-pentafluorosulfanyl phenol, 4-hydroxyphenylsulfur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl phenol, 4-pentafluoro-lambda∼6∼-sulfanyl phenol, sulfur,pentafluoro 4-hydroxyphenyl-, oc-6-21-9ci, 4-pentafluorosulfanylphenol, 4-pentafluorosulfur phenol, 4-pentafluorosulphanyl phenol, 4-pentafluorothio vi phenol
PubChem CID 2779203
IUPAC Name 4-(pentafluoro-$l^{6}-sulfanyl)phenol
SMILES OC1=CC=C(C=C1)S(F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3341303
View Documents
Thermo Scientific Chemicals
H3341303
1 g
Each for $201.25
Only null left
 
AAH3341306
View Documents
Thermo Scientific Chemicals
H3341306
5 g
Each for $792.62
Only null left
 
Description

Description

Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

774-94-7
220.16
XHJLGVIUMCBMHL-UHFFFAOYSA-N
2779203
OC1=CC=C(C=C1)S(F)(F)(F)(F)F
C6H5F5OS
MFCD03788516
4-pentafluorothio phenol, 4-pentafluorosulfanyl phenol, 4-hydroxyphenylsulfur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl phenol, 4-pentafluoro-lambda∼6∼-sulfanyl phenol, sulfur,pentafluoro 4-hydroxyphenyl-, oc-6-21-9ci, 4-pentafluorosulfanylphenol, 4-pentafluorosulfur phenol, 4-pentafluorosulphanyl phenol, 4-pentafluorothio vi phenol
4-(pentafluoro-$l^{6}-sulfanyl)phenol

Specifications

774-94-7
MFCD03788516
4-pentafluorothio phenol, 4-pentafluorosulfanyl phenol, 4-hydroxyphenylsulfur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl phenol, 4-pentafluoro-lambda∼6∼-sulfanyl phenol, sulfur,pentafluoro 4-hydroxyphenyl-, oc-6-21-9ci, 4-pentafluorosulfanylphenol, 4-pentafluorosulfur phenol, 4-pentafluorosulphanyl phenol, 4-pentafluorothio vi phenol
OC1=CC=C(C=C1)S(F)(F)(F)(F)F
220.16
220.16
4-(Pentafluorothio)phenol
C6H5F5OS
1 g
XHJLGVIUMCBMHL-UHFFFAOYSA-N
4-(pentafluoro-$l^{6}-sulfanyl)phenol
2779203
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only