Learn More
4-Oxo-L-proline hydrobromide, 90+%
CAS: 75776-67-9 | C5H8BrNO3 | 210.027 g/mol
$80.49 - $206.53
Chemical Identifiers
CAS | 75776-67-9 |
---|---|
Molecular Formula | C5H8BrNO3 |
Molecular Weight (g/mol) | 210.027 |
MDL Number | MFCD12024572 |
InChI Key | SFSROLZHCPCODJ-WCCKRBBISA-N |
Synonym | 4-keto-l-proline hydrobromide, s-4-oxopyrrolidine-2-carboxylic acid hydrobromide, 4-keto-l-prolinehydrobromide, 4-oxo-l-proline hydrobromide, 4-oxoproline hydrobromide, l-proline, 4-oxo-, hydrobromide, s-4-keto-proline hydrobromide, pubchem14289, 4-keto-l-proline, hydrobromide |
PubChem CID | 12773661 |
IUPAC Name | (2S)-4-oxopyrrolidine-2-carboxylic acid;hydrobromide |
SMILES | C1C(NCC1=O)C(=O)O.Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH57691MD
|
Thermo Scientific Chemicals
H57691MD |
250 mg |
Each for $80.49
|
|
|||||
AAH5769103
|
Thermo Scientific Chemicals
H5769103 |
1 g |
Each for $206.53
|
|
|||||
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
75776-67-9 | |
210.027 | |
SFSROLZHCPCODJ-WCCKRBBISA-N | |
12773661 | |
C1C(NCC1=O)C(=O)O.Br |
C5H8BrNO3 | |
MFCD12024572 | |
4-keto-l-proline hydrobromide, s-4-oxopyrrolidine-2-carboxylic acid hydrobromide, 4-keto-l-prolinehydrobromide, 4-oxo-l-proline hydrobromide, 4-oxoproline hydrobromide, l-proline, 4-oxo-, hydrobromide, s-4-keto-proline hydrobromide, pubchem14289, 4-keto-l-proline, hydrobromide | |
(2S)-4-oxopyrrolidine-2-carboxylic acid;hydrobromide |
Specifications
134°C to 140°C | |
C5H8BrNO3 | |
4-keto-l-proline hydrobromide, s-4-oxopyrrolidine-2-carboxylic acid hydrobromide, 4-keto-l-prolinehydrobromide, 4-oxo-l-proline hydrobromide, 4-oxoproline hydrobromide, l-proline, 4-oxo-, hydrobromide, s-4-keto-proline hydrobromide, pubchem14289, 4-keto-l-proline, hydrobromide | |
C1C(NCC1=O)C(=O)O.Br | |
250 mg | |
12773661 | |
210.03 | |
≥90% |
75776-67-9 | |
MFCD12024572 | |
SFSROLZHCPCODJ-WCCKRBBISA-N | |
(2S)-4-oxopyrrolidine-2-carboxylic acid;hydrobromide | |
210.027 | |
−32° (c=0.5 in Methanol) | |
Moisture sensitive | |
4-Oxo-L-proline hydrobromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only