missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M127520LTMT
Specifications
108-10-1 | |
Steel Can | |
20 L | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
4-methylpentan-2-one | |
0.985 | |
0.002 meq/g |
1 | |
C6H12O | |
5e-05 | |
CC(C)CC(C)=O | |
100.16 | |
ACS |
Chemical Identifiers
108-10-1 | |
100.16 | |
4-methylpentan-2-one |
C6H12O | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
CC(C)CC(C)=O |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.