missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, ACS, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M12751LT

 View more versions of this product

Catalog No. 18608271


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

108-10-1
Amber Glass Bottle
1 L
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
0.985
0.002 meq/g
1
C6H12O
5e-05
CC(C)CC(C)=O
100.16
ACS

Chemical Identifiers

108-10-1
100.16
4-methylpentan-2-one
C6H12O
NTIZESTWPVYFNL-UHFFFAOYSA-N
CC(C)CC(C)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.