missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl 4-O-Acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside 98.0+%, TCI America™
$57.79 - $298.25
Chemical Identifiers
CAS | 140615-77-6 |
---|---|
Molecular Formula | C37H35NO9 |
Molecular Weight (g/mol) | 637.685 |
InChI Key | QZTMIUALKXAHCQ-RHPHZSOCSA-N |
Synonym | GlcNPhth[36Bn,4Ac]-beta-MP |
PubChem CID | 133554185 |
IUPAC Name | [(2R,3R,4S,5R,6R)-5-(1,3-dioxoisoindol-2-yl)-6-(4-methoxyphenoxy)-4-phenylmethoxy-2-(phenylmethoxymethyl)oxan-3-yl] acetate |
SMILES | CC(=O)OC1C(OC(C(C1OCC2=CC=CC=C2)N3C(=O)C4=CC=CC=C4C3=O)OC5=CC=C(C=C5)OC)COCC6=CC=CC=C6 |
Chemical Identifiers
140615-77-6 | |
637.685 | |
GlcNPhth[36Bn,4Ac]-beta-MP | |
[(2R,3R,4S,5R,6R)-5-(1,3-dioxoisoindol-2-yl)-6-(4-methoxyphenoxy)-4-phenylmethoxy-2-(phenylmethoxymethyl)oxan-3-yl] acetate |
C37H35NO9 | |
QZTMIUALKXAHCQ-RHPHZSOCSA-N | |
133554185 | |
CC(=O)OC1C(OC(C(C1OCC2=CC=CC=C2)N3C(=O)C4=CC=CC=C4C3=O)OC5=CC=C(C=C5)OC)COCC6=CC=CC=C6 |
Specifications
140615-77-6 | |
C37H35NO9 | |
GlcNPhth[36Bn,4Ac]-beta-MP | |
CC(=O)OC1C(OC(C(C1OCC2=CC=CC=C2)N3C(=O)C4=CC=CC=C4C3=O)OC5=CC=C(C=C5)OC)COCC6=CC=CC=C6 | |
637.685 | |
637.69 | |
Crystalline Powder |
White | |
1 g | |
QZTMIUALKXAHCQ-RHPHZSOCSA-N | |
[(2R,3R,4S,5R,6R)-5-(1,3-dioxoisoindol-2-yl)-6-(4-methoxyphenoxy)-4-phenylmethoxy-2-(phenylmethoxymethyl)oxan-3-yl] acetate | |
133554185 | |
≥98.0% (HPLC) | |
4-Methoxyphenyl 4-O-Acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator